missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethoxy-2-propanol, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 259050010

 View more versions of this product

Catalog No. AC259050010



Description

Description

Specifications

Specifications

1-Ethoxy-2-propanol
42°C
Glass Bottle
0.897
0.1% Max. (K.F.)
Colorless
1L
94% min. Remark: balance 2-ethoxy-1-propanol (GC)
C2H5OCH2CH(OH)CH3
Solubility in water: soluble
CCOCC(C)O
104.14
104.14
95%
0.8970g/mL
Authentic
1.4050 to 1.409
2 mPa.s (20°C)
132°C
-100°C
1569-02-4
C5H12O2
1-ethoxy-2-propan, 1-ethoxy-2-propano, 1-ethoxy-2-propanol, 1-ethoxy-propan-2-ol, 2-hydroxypropylethylether, 2-propanol, 1-ethoxy, acmc-1byqk, arcosolv r pe, propylene glycol ethyl ether, propylene glycol monoethyl ether
JOLQKTGDSGKSKJ-UHFFFAOYSA-N
1-ethoxypropan-2-ol
15287
Liquid

Chemical Identifiers

1569-02-4
104.14
1-ethoxy-2-propan, 1-ethoxy-2-propano, 1-ethoxy-2-propanol, 1-ethoxy-propan-2-ol, 2-hydroxypropylethylether, 2-propanol, 1-ethoxy, acmc-1byqk, arcosolv r pe, propylene glycol ethyl ether, propylene glycol monoethyl ether
1-ethoxypropan-2-ol
C5H12O2
JOLQKTGDSGKSKJ-UHFFFAOYSA-N
15287
CCOCC(C)O
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 216-374-5

SDS Documents