missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Ethoxy-2-propanol, 90+%, remainder 2-ethoxy-1-propanol

Manufacturer:  Alfa Aesar™ 43306AY

 View more versions of this product

Catalog No. AA43306AY



Specifications

Specifications

1-Ethoxy-2-propanol
42°C (108°F)
132°C
-100°C
1569-02-4
C5H12O2
MFCD00067050
1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
CCOCC(C)O
104.149
104.15
≥90%
0.896g/mL
1.4065
Colorless
4 x 500mL
Remainder 2-ethoxy-1-propanol
CH3CH2OCH2CH(OH)CH3
UN1987
JOLQKTGDSGKSKJ-UHFFFAOYSA-N
1-ethoxypropan-2-ol
15287
Liquid

Chemical Identifiers

1569-02-4
104.149
JOLQKTGDSGKSKJ-UHFFFAOYSA-N
15287
CCOCC(C)O
C5H12O2
MFCD00067050
1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
1-ethoxypropan-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P280-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

Documents