missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene, TCI America™

Manufacturer:  TCI America E12305G

 View more versions of this product

Catalog No. E12305G



Specifications

Specifications

1-Ethoxy-2,3-difluoro-4-(trans-4-propylcyclohexyl)benzene
5g
C17H24F2O
BOAHGIPRRKDVQY-UHFFFAOYSA-N
1-ethoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene
19103501
Crystal-Powder at 20°C
White-Yellow
174350-05-1
MFCD11053400
CCCC1CCC(CC1)C2=C(C(=C(C=C2)OCC)F)F
282.375
282.37
≥98.0% (GC)

Chemical Identifiers

174350-05-1
282.375
BOAHGIPRRKDVQY-UHFFFAOYSA-N
1-ethoxy-2,3-difluoro-4-(4-propylcyclohexyl)benzene
C17H24F2O
MFCD11053400
19103501
CCCC1CCC(CC1)C2=C(C(=C(C=C2)OCC)F)F
Safety and Handling

Safety and Handling

EINECSNumber : (4)-1953

TSCA : No

Documents