Please login to your online account to display your discounted pricing

1-Dodecyne, 98%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC223450250

 View more versions of this product

Catalog No. AC223450250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 166.31
Physical Form Liquid
Quantity 25mL
Color Clear colorless to light yellow
Density 0.77g/mL
Beilstein 01, 261
CAS Max % 100.0
Refractive Index 1.4330 to 1.4350 (20°C, 589nm)
Molecular Formula C12H22
CAS 765-03-7
Chemical Name or Material 1-Dodecyne
Assay Percent Range 98%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00008960
CAS Min % 97.5
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula CH3(CH2)9C2CH
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 1-Dodecyne
CAS 765-03-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H22
Linear Formula CH3(CH2)9C2CH
Formula Weight 166.31
MDL Number MFCD00008960
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents