Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dodecyne, 97%

$43.73 - $146.69

Chemical Identifiers

Chemical Name or Material 1-Dodecyne
CAS 765-03-7
Molecular Formula CH3(CH2)9C≡CH
Formula Weight 166.31
MDL Number MFCD00008960
Beilstein 1741320
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0297706

 
alfa aesar™
L0297706
5g Each for $43.73

AAL0297714

 
alfa aesar™
L0297714
25g Each for $146.69
Description & Specifications

Specifications

Formula Weight 166.31
Chemical Name or Material 1-Dodecyne
Assay 97%
Molecular Formula CH3(CH2)9C≡CH
CAS 765-03-7
Health Hazard 1 Warning
TSCA TSCA
Beilstein 1741320
Boiling Point 214° to 216°C
Melting Point −19°C
Flash Point 79°C (174°F)
MDL Number MFCD00008960
Quantity 5g

Applications
1-Dodecyne was used in the synthesis of stable ruthenium nanoparticles.

Solubility
Not miscible in water.

Notes
Incompatible with heavy metal powders and oxidizing agents. Store between 2 - 8°C. Store away from oxidizing agents.

Safety and Handling

Warning