Learn More
1-Dodecylphosphonic acid, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2625909
Description
1-Dodecylphosphonic acid is used as tuning of surface energy, wetting, and work function of metal oxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Dodecylphosphonic acid is used as tuning of surface energy, wetting, and work function of metal oxides.
Solubility
Sparingly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents, bases.
Specifications
1-Dodecylphosphonic acid | |
92°C to 96°C | |
MFCD00015832 | |
UN3261 | |
Sparingly soluble in water. | |
CCCCCCCCCCCCP(O)(O)=O | |
250.32 | |
250.31 |
5137-70-2 | |
C12H27O3P | |
10 g | |
n-dodecylphosphonic acid, phosphonic acid, dodecyl, n-dodecanephosphonic acid, dodecane phosphonic acid, 1-dodecanephosphonic acid, n-dodecyl phosphonic acid, dodecyl-phosphonic acid, 1-dodecylphosphonic acid, phosphonic acid, p-dodecyl, n-dodecylphosphonicacid | |
SVMUEEINWGBIPD-UHFFFAOYSA-N | |
dodecylphosphonic acid | |
78816 | |
95% |
Chemical Identifiers
5137-70-2 | |
250.32 | |
SVMUEEINWGBIPD-UHFFFAOYSA-N | |
78816 | |
CCCCCCCCCCCCP(O)(O)=O |
C12H27O3P | |
MFCD00015832 | |
n-dodecylphosphonic acid, phosphonic acid, dodecyl, n-dodecanephosphonic acid, dodecane phosphonic acid, 1-dodecanephosphonic acid, n-dodecyl phosphonic acid, dodecyl-phosphonic acid, 1-dodecylphosphonic acid, phosphonic acid, p-dodecyl, n-dodecylphosphonicacid | |
dodecylphosphonic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335-H373
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 225-897-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only