Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dodecylmagnesium bromide, 0.5M in MeTHF

Chemical Identifiers

Chemical Name or Material 1-Dodecylmagnesium bromide
CAS 15890-72-9
Molecular Formula C12H25BrMg
Linear Formula CH3(CH2)11MgBr
Formula Weight 273.54
MDL Number MFCD00075165
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51172AP

  SDS
alfa aesar™
H51172AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 273.54
Concentration 0.5M in MeTHF
Health Hazard 2 GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
CAS 15890-72-9
Quantity 500mL
Health Hazard 1 Danger
Linear Formula CH3(CH2)11MgBr
MDL Number MFCD00075165
Health Hazard 3 GHS P Statement
P210-P280-P305+P351+P338-P309-P310-P402
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.
Molecular Formula C12H25BrMg
UN Number UN2924
Chemical Name or Material 1-Dodecylmagnesium bromide