Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dodecylboronic acid pinacol ester, 98%

$89.04 - $982.41

Chemical Identifiers

Chemical Name or Material 1-Dodecylboronic acid pinacol ester
CAS 177035-82-4
Molecular Formula C18H37BO2
Formula Weight 296.31
MDL Number MFCD12546195
Synonym 2-n-Dodecyl-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3118603

  SDS
alfa aesar™
H3118603
1g Each for $89.04

AAH3118606

  SDS
alfa aesar™
H3118606
5g Each for $296.84

AAH3118614

  SDS
alfa aesar™
H3118614
25g Each for $982.41
Description & Specifications

Specifications

Formula Weight 296.31
Chemical Name or Material 1-Dodecylboronic acid pinacol ester
Assay 98%
Molecular Formula C18H37BO2
MDL Number MFCD12546195
CAS 177035-82-4
Synonym 2-n-Dodecyl-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
Boiling Point 123°C (0.2mmHg)
Quantity 1g