Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dodecylboronic acid, 98%

$58.81 - $677.28

Chemical Identifiers

Chemical Name or Material 1-Dodecylboronic acid
CAS 3088-79-7
Molecular Formula C12H27BO2
Linear Formula CH3(CH2)11B(OH)2
Formula Weight 214.16
MDL Number MFCD04039322
Beilstein 1750795
Synonym n-Dodecaneboronic acid; Laurylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1995803

 
alfa aesar™
L1995803
1g Each for $58.81

AAL1995806

 
alfa aesar™
L1995806
5g Each for $173.93

AAL1995814

 
alfa aesar™
L1995814
25g Each for $677.28
Description & Specifications

Specifications

Formula Weight 214.16
Chemical Name or Material 1-Dodecylboronic acid
Molecular Formula C12H27BO2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
MDL Number MFCD04039322
CAS 3088-79-7
Beilstein 1750795
Synonym n-Dodecaneboronic acid; Laurylboronic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Linear Formula CH3(CH2)11B(OH)2
Melting Point 91° to 93°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.