missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dodecylamine hydrochloride, 97%

Manufacturer:  Alfa Aesar™ L1231418

 View more versions of this product

Catalog No. AAL1231418



Specifications

Specifications

1-Dodecylamine hydrochloride
50g
C12H28ClN
MFCD00042017
dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd
CCCCCCCCCCCCN.Cl
221.813
221.82
186°C to 188°C
929-73-7
CH3(CH2)11NH2·HCl
3553943
TWFQJFPTTMIETC-UHFFFAOYSA-N
dodecan-1-amine;hydrochloride
458426
97%

Chemical Identifiers

929-73-7
221.813
TWFQJFPTTMIETC-UHFFFAOYSA-N
458426
CCCCCCCCCCCCN.Cl
C12H28ClN
MFCD00042017
dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd
dodecan-1-amine;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : JR6750000

TSCA : TSCA

Documents