missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Dodecylamine, 98+%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1551522

 View more versions of this product

Catalog No. AAA1551522



Description

Description

Applications
Dodecylamine was used in preparation of novel surfactant copper(II) complexes. It is used as a catalyst and template agent in the sol-gel process for the fabrication of monodispersed mesoporous bioactive glass sub-micron spheres.

Solubility
Soluble in water (78 mg/L 25°C).

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications

Specifications

1-Dodecylamine
100g
≥98%
MFCD00008154
1633576
JRBPAEWTRLWTQC-UHFFFAOYSA-N
dodecan-1-amine
13583
≥98%
0.802
124-22-1
C12H27N
UN3259
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4
CCCCCCCCCCCCN
185.355
185.36

Chemical Identifiers

124-22-1
185.355
JRBPAEWTRLWTQC-UHFFFAOYSA-N
13583
CCCCCCCCCCCCN
C12H27N
MFCD00008154
dodecylamine, 1-dodecanamine, laurylamine, 1-aminododecane, n-dodecylamine, dodecanamine, 1-dodecylamine, lauramine, n-laurylamine, alamine 4
dodecan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.

RTECSNumber : JR6475000

TSCA : TSCA

Documents