Learn More
(1-Dodecyl)trimethylammonium bromide, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1076118
Description
(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.
Specifications
(1-Dodecyl)trimethylammonium bromide | |
∼245°C (decomposition) | |
MFCD00011767 | |
UN3077 | |
Hygroscopic | |
Soluble in water. | |
CCCCCCCCCCCC[N+](C)(C)C.[Br-] | |
308.348 | |
CHEBI:282662 | |
99% |
1119-94-4 | |
C15H34BrN | |
50 g | |
3597463 | |
dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide | |
XJWSAJYUBXQQDR-UHFFFAOYSA-M | |
dodecyl(trimethyl)azanium;bromide | |
14249 | |
308.35 |
Chemical Identifiers
1119-94-4 | |
308.348 | |
XJWSAJYUBXQQDR-UHFFFAOYSA-M | |
14249 | |
dodecyl(trimethyl)azanium;bromide |
C15H34BrN | |
MFCD00011767 | |
dodecyltrimethylammonium bromide, dtab, dctab, ltab, lauryltrimethylammonium bromide, n,n,n-trimethyldodecan-1-aminium bromide, laurtrimonium bromide, dodecyl trimethyl ammonium bromide, morpan d, 1-dodecanaminium, n,n,n-trimethyl-, bromide | |
CHEBI:282662 | |
CCCCCCCCCCCC[N+](C)(C)C.[Br-] |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 214-290-3
RTECSNumber : BQ3195000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only