Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dodecyl isothiocyanate, tech. 90%

Chemical Identifiers

Chemical Name or Material 1-Dodecyl isothiocyanate
CAS 1072-32-8
Purity Grade Technical Grade
Molecular Formula CH3(CH2)11NCS
Formula Weight 227.41
MDL Number MFCD00041121
UN Number UN1760
Beilstein 1774889
Synonym Lauryl isothiocyanate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2180203

  SDS
alfa aesar™
B2180203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2180206

  SDS
alfa aesar™
B2180206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 227.41
Odor Irritating
Purity Grade Technical Grade
Quantity 1g
Health Hazard 1 Danger
Beilstein 1774889
Boiling Point 173° to 174°C (1mmHg)
Molecular Formula CH3(CH2)11NCS
Health Hazard 2 GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Assay 90%
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
MDL Number MFCD00041121
CAS 1072-32-8
Synonym Lauryl isothiocyanate
Chemical Name or Material 1-Dodecyl isothiocyanate
UN Number UN1760
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.