missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dodecyl isocyanate, 98%

Manufacturer:  Alfa Aesar™ L1338006

 View more versions of this product

Catalog No. AAL1338006



Description

Description

Applications
1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

Solubility
It hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

1-Dodecyl isocyanate
Irritating
5g
C13H25NO
UN2206
dodecyl isocyanate, dodecane, 1-isocyanato, dodecylisocyanate, 1-dodecyl isocyanate, n-dodecyl isocyanate, acmc-209jmx, dodecyl isocyanate, isocyanic acid dodecyl ester, 1-dodecyl isocyanate
CCCCCCCCCCCCN=C=O
211.349
211.35
134°C (273°F)
157°C to 161°C (21mmHg)
4202-38-4
MFCD00041744
1774823
YIDSTEJLDQMWBR-UHFFFAOYSA-N
1-isocyanatododecane
77873
98%

Chemical Identifiers

4202-38-4
211.349
YIDSTEJLDQMWBR-UHFFFAOYSA-N
77873
CCCCCCCCCCCCN=C=O
C13H25NO
MFCD00041744
dodecyl isocyanate, dodecane, 1-isocyanato, dodecylisocyanate, 1-dodecyl isocyanate, n-dodecyl isocyanate, acmc-209jmx, dodecyl isocyanate, isocyanic acid dodecyl ester, 1-dodecyl isocyanate
1-isocyanatododecane
Safety and Handling

Safety and Handling

GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents