Learn More
1-Dodecene, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1499222
Description
1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.
Solubility
It is soluble in ethanol, ethyl ether, and acetone
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.
Specifications
1-Dodecene | |
-37°C | |
212°C to 213°C | |
C12H24 | |
CH3(CH2)9CH=CH2 | |
100 g | |
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr | |
CRSBERNSMYQZNG-UHFFFAOYSA-N | |
dodec-1-ene | |
8183 | |
96% |
112-41-4 | |
0.758 | |
73°C (163°F) | |
1.43 | |
MFCD00008961 | |
1699848 | |
It is soluble in ethanol,ethyl ether,and acetone | |
CCCCCCCCCCC=C | |
168.324 | |
168.32 |
Chemical Identifiers
112-41-4 | |
168.324 | |
CRSBERNSMYQZNG-UHFFFAOYSA-N | |
8183 | |
CCCCCCCCCCC=C |
C12H24 | |
MFCD00008961 | |
1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr | |
dodec-1-ene |
Safety and Handling
GHS H Statement
H304-H315-H227
May be fatal if swallowed and enters airways.
Causes skin irritation.
Combustible liquid.
P210-P280-P301+P310-P331-P370+P378q-P501c
H227-H304
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 203-968-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only