Learn More
1-Dodecanol, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 155450050
Specifications
1-Dodecanol | |
119°C | |
<0.1mbar at 20°C | |
Colorless to Yellow | |
5mL | |
97.5 | |
98% | |
CH3(CH2)11OH | |
01, 428 | |
1-dodecanol, 1-dodecyl alcohol, dodecanol, dodecyl alcohol, dodecylalcohol, lauric alcohol, laurinic alcohol, lauryl alcohol, n-dodecyl alcohol, undecyl carbinol | |
LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
dodecan-1-ol | |
8193 | |
186.34 | |
98% |
0.8350g/mL | |
Authentic | |
258.0°C to 265.0°C | |
22.0°C to 27.0°C | |
112-53-8 | |
100.0 | |
C12H26O | |
MFCD00004753 | |
15, 3451 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CCCCCCCCCCCCO | |
186.339 | |
CHEBI:28878 | |
Liquid |
Chemical Identifiers
112-53-8 | |
186.339 | |
LQZZUXJYWNFBMV-UHFFFAOYSA-N | |
8193 | |
dodecan-1-ol |
C12H26O | |
MFCD00004753 | |
1-dodecanol, 1-dodecyl alcohol, dodecanol, dodecyl alcohol, dodecylalcohol, lauric alcohol, laurinic alcohol, lauryl alcohol, n-dodecyl alcohol, undecyl carbinol | |
CHEBI:28878 | |
CCCCCCCCCCCCO |
Safety and Handling
GHS P Statement
Causes serious eye irritation.
Very toxic to aquatic life.
GHS P Statement
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning