Please login to your online account to display your discounted pricing

1-Dodecanesulfonic Acid, Sodium Salt, Monohydrate, HPLC Grade, 99% min., EMD Millipore


Quantity 100g

Manufacturer: emd millipore corporation  DX01023

 View more versions of this product

Catalog No. MDX01023

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Formula Weight 272.37
Color White
Quantity 100g
Bulk Density 330kg/m3
Chemical Name or Material 1-Dodecanesulfonic Acid, Sodium Salt, Monohydrate
Name Note LiChropur™, For Ion Pair Chromatography
Assay Percent Range 99% min.
Molecular Formula CH3(CH2)11SO3Na·H2O
Linear Formula C12H25NaO3S
CAS 2386-53-0
Synonym Sodium laurylsulfonate
Recommended Storage Store at +5°C to +30°C

Specifications

Chemical Name or Material 1-Dodecanesulfonic Acid, Sodium Salt, Monohydrate
Name Note LiChropur™, For Ion Pair Chromatography
CAS 2386-53-0
Physical Form Crystalline Solid
Assay Percent Range 99% min.
Molecular Formula CH3(CH2)11SO3Na·H2O
Linear Formula C12H25NaO3S
Formula Weight 272.37
Synonym Sodium laurylsulfonate
Accessories

Store at +5°C to +30°C