Please login to your online account to display your discounted pricing

1-Dodecanesulfonic Acid, Sodium Salt, Monohydrate, HPLC Grade, 99% min., EMD Millipore

$371.09 - $1,090.63

Chemical Identifiers

Chemical Name or Material 1-Dodecanesulfonic Acid, Sodium Salt, Monohydrate
Name Note LiChropur™, For Ion Pair Chromatography
CAS 2386-53-0
Physical Form Crystalline Solid
Assay Percent Range 99% min.
Molecular Formula CH3(CH2)11SO3Na·H2O
Linear Formula C12H25NaO3S
Formula Weight 272.37
Synonym Sodium laurylsulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MDX01023

 
emd millipore corporation
DX01023
100g Each for $1,090.63

MDX01022

 
emd millipore corporation
DX01022
25g Each for $371.09
Description & Specifications

Specifications

Physical Form Crystalline Solid
Formula Weight 272.37
Color White
Quantity 100g
Bulk Density 330kg/m3
Linear Formula C12H25NaO3S
CAS 2386-53-0
Name Note LiChropur™, For Ion Pair Chromatography
Molecular Formula CH3(CH2)11SO3Na·H2O
Assay Percent Range 99% min.
Chemical Name or Material 1-Dodecanesulfonic Acid, Sodium Salt, Monohydrate
Synonym Sodium laurylsulfonate
Recommended Storage Store at +5°C to +30°C
Safety and Handling

Store at +5°C to +30°C