missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Dodecanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC396720050

 View more versions of this product

Catalog No. AC396720050


 

Specifications Description & Specifications

Specifications

0.02 max. (C=0.18 M in hot water) at 230nm
5g
2386-53-0
Membrane filtration test:Passes Test on 0.18M sol
2% max.
Authentic
≥99.0% (alkal. after ion exchange) on dry substance
C12H25NaO3S
CH3(CH2)11SO3Na
MFCD00007527
White
1-Dodecanesulfonic acid, sodium salt
Anhydrous, Ion pair chromatography
99.0
100.0
Ion pair chromatography
Crystalline Powder
99+%
Glass bottle
272.38
HPLC gradient (Passes)

Chemical Identifiers

2386-53-0
100.0
C12H25NaO3S
272.38
1-Dodecanesulfonic acid, sodium salt, Anhydrous, Ion pair chromatography
Ion pair chromatography
99.0
99+%
CH3(CH2)11SO3Na
MFCD00007527
≥99.0% (alkal. after ion exchange) on dry substance