Please login to your online account to display your discounted pricing

1-(Diphenylmethyl)piperazine 97%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  122931000

 View more versions of this product

Catalog No. AC122931000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0% Total Base (on dry substance), ≥98.0% (TLC)
Formula Weight 252.36
Quantity 100g
Color White to light yellow
EINECS Number 212-667-7
TSCA TSCA
Melting Point 88° to 93°C
RTECS Number TL6465000
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 1-(Diphenylmethyl)piperazine
Infrared Spectrum Authentic
Name Note 97%
Packaging Plastic bottle
Assay Percent Range 97%
Molecular Formula C17H20N2
MDL Number MFCD00038379
CAS 841-77-0
Water 2% max. (K.F.)
Synonym Benzhydrylpiperazine
Health Hazard 1 AIR SENSITIVE
Health Hazard 2 GHS P Statement Harmful if swallowed.
Health Hazard 3 GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Specifications

Chemical Name or Material 1-(Diphenylmethyl)piperazine
Name Note 97%
CAS 841-77-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% Total Base (on dry substance), ≥98.0% (TLC)
Assay Percent Range 97%
Molecular Formula C17H20N2
Formula Weight 252.36
MDL Number MFCD00038379
Synonym Benzhydrylpiperazine
Accessories

AIR SENSITIVE

GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS P Statement Harmful if swallowed.

SDS & Safety Documents