Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt, 95%

$27.27 - $406.82

Chemical Identifiers

Chemical Name or Material 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt
CAS 136997-71-2
Molecular Formula C10H14N2O4S
Formula Weight 258.29
MDL Number MFCD00185917
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2212409

  SDS
alfa aesar™
B2212409
10g Each for $27.27

AAB2212418

  SDS
alfa aesar™
B2212418
50g Each for $100.96

AAB2212430

  SDS
alfa aesar™
B2212430
250g Each for $406.82
Description & Specifications

Specifications

Formula Weight 258.29
CAS 136997-71-2
Health Hazard 3 GHS P Statement
P261-P280g-P262
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.
Chemical Name or Material 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt
TSCA TSCA
Assay 95%
Molecular Formula C10H14N2O4S
Melting Point ≈180°C (dec.)
MDL Number MFCD00185917
Quantity 10g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Safety and Handling

Warning

GHS P Statement
P261-P280g-P262
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.

GHS H Statement
H317
May cause an allergic skin reaction.