Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Dimethylaminomethyl)cyclopentanecarboxaldehyde, 97%

$94.47 - $323.96

Chemical Identifiers

Chemical Name or Material 1-(Dimethylaminomethyl)cyclopentanecarboxaldehyde
CAS 39943-37-8
Molecular Formula C9H17NO
Formula Weight 155.24
MDL Number MFCD08064242
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5075103

  SDS
alfa aesar™
H5075103
1g Each for $323.96

AAH50751MD

  SDS
alfa aesar™
H50751MD
250mg Each for $94.47
Description & Specifications

Specifications

Formula Weight 155.24
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-(Dimethylaminomethyl)cyclopentanecarboxaldehyde
MDL Number MFCD08064242
Health Hazard 1 Warning
CAS 39943-37-8
Assay 97%
Molecular Formula C9H17NO
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.