Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dimethylaminomethyl-2-naphthol, 98%

$67.66 - $241.08

Chemical Identifiers

Chemical Name or Material 1-Dimethylaminomethyl-2-naphthol
CAS 5419-02-3
Molecular Formula C13H15NO
Formula Weight 201.27
MDL Number MFCD00086150
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5043003

  SDS
alfa aesar™
H5043003
1g Each for $241.08

AAH50430MD

  SDS
alfa aesar™
H50430MD
250mg Each for $67.66
Description & Specifications

Specifications

Formula Weight 201.27
Quantity 1g
CAS 5419-02-3
Molecular Formula C13H15NO
Assay 98%
MDL Number MFCD00086150
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 1-Dimethylaminomethyl-2-naphthol
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.