Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Dimethylamino-2-pentyne, 98%

Chemical Identifiers

Chemical Name or Material 1-Dimethylamino-2-pentyne
CAS 7383-77-9
Molecular Formula C7H13N
Linear Formula CH3CH2C≡CCH2N(CH3)2
Formula Weight 111.19
MDL Number MFCD00041631
UN Number UN2734
Beilstein 1738713
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0566206

  SDS
alfa aesar™
L0566206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0566214

  SDS
alfa aesar™
L0566214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 111.19
Chemical Name or Material 1-Dimethylamino-2-pentyne
Assay 98%
MDL Number MFCD00041631
CAS 7383-77-9
UN Number UN2734
Beilstein 1738713
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
Molecular Formula C7H13N
Linear Formula CH3CH2C≡CCH2N(CH3)2
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.