Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Difluoromethoxy-4-nitrobenzene, 98%

$96.97 - $357.11

Chemical Identifiers

Chemical Name or Material 1-Difluoromethoxy-4-nitrobenzene
CAS 1544-86-1
Molecular Formula C7H5F2NO3
Formula Weight 189.12
MDL Number MFCD00085009
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2191006

 
alfa aesar™
B2191006
5g Each for $96.97

AAB2191014

 
alfa aesar™
B2191014
25g Each for $357.11
Description & Specifications

Specifications

Formula Weight 189.12
Chemical Name or Material 1-Difluoromethoxy-4-nitrobenzene
Assay 98%
Molecular Formula C7H5F2NO3
MDL Number MFCD00085009
CAS 1544-86-1
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Characteristic
Boiling Point 108° to 110°C (1.5mmHg)
Melting Point 33° to 35°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.