Please login to your online account to display your discounted pricing

1-Deoxynojirimycin, Tocris

Glucosidase I and II inhibitor

$175.83 - $671.19

Chemical Identifiers

Chemical Name or Material 1-Deoxynojirimycin
Assay Percent Range >98%
Molecular Formula C6H13NO4
Formula Weight 163.17
Synonym (2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

125825

 
r & d systems
RDS125825
25mg Each for $671.19

12585

  SDS
r & d systems
RDS12585
5mg Each for $175.83
Description & Specifications

Specifications

Formula Weight 163.17
Quantity 25mg
Chemical Name or Material 1-Deoxynojirimycin
Synonym (2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol
CAS 19130-96-2
Assay Percent Range >98%
Molecular Formula C6H13NO4
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C