Please login to your online account to display your discounted pricing

1-Deoxynojirimycin, Tocris


Glucosidase I and II inhibitor

Quantity 5mg

Manufacturer: r & d systems  RDS12585

 View more versions of this product

Catalog No. 12585

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 163.17
Quantity 5mg
Chemical Name or Material 1-Deoxynojirimycin
Molecular Formula C6H13NO4
CAS 19130-96-2
Synonym (2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol
Assay Percent Range >98%
Recommended Storage Store at +4°C

Specifications

Chemical Name or Material 1-Deoxynojirimycin
Assay Percent Range >98%
Molecular Formula C6H13NO4
Formula Weight 163.17
Synonym (2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol
Accessories

Store at +4°C