missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (+)-1-Deoxymannojirimycin hydrochloride

Selective inhibitor of a-mannosidase

$206.65 - $595.26

Chemical Identifiers

CAS 73465-43-7
Molecular Formula C6H14ClNO4
Molecular Weight (g/mol) 199.631
MDL Number MFCD00083611
InChI Key ZJIHMALTJRDNQI-MVNLRXSJSA-N
Synonym DMJ; 1,5-Dideoxy-1,5-imino-D-mannitol hydrochloride
PubChem CID 11390018
IUPAC Name (2R,3R,4R,5R)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES C1C(C(C(C(N1)CO)O)O)O.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ61277LB0 View Documents Alfa Aesar™
J61277LB0
5mg Each for $206.65
 
AAJ61277MA View Documents Alfa Aesar™
J61277MA
10mg Each for $343.89
 
AAJ61277MB View Documents Alfa Aesar™
J61277MB
25mg Each for $595.26
 
Specifications

Chemical Identifiers

73465-43-7
199.631
ZJIHMALTJRDNQI-MVNLRXSJSA-N
11390018
C1C(C(C(C(N1)CO)O)O)O.Cl
C6H14ClNO4
MFCD00083611
DMJ; 1,5-Dideoxy-1,5-imino-D-mannitol hydrochloride
(2R,3R,4R,5R)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride

Specifications

73465-43-7
MFCD00083611
ZJIHMALTJRDNQI-MVNLRXSJSA-N
(2R,3R,4R,5R)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
11390018
Powder
5mg
C6H14ClNO4
DMJ; 1,5-Dideoxy-1,5-imino-D-mannitol hydrochloride
C1C(C(C(C(N1)CO)O)O)O.Cl
199.631
199.6
White
(+)-1-Deoxymannojirimycin hydrochloride
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C