Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Deoxy-1-nitro-L-iditol hemihydrate, 99%

$237.05 - $237.05

Chemical Identifiers

Chemical Name or Material 1-Deoxy-1-nitro-L-iditol hemihydrate
CAS 207226-23-1
Molecular Formula C6H13NO7·0.5H2O
Formula Weight 220.19 (211.18 Anhydrous)
MDL Number MFCD00191972
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB21616ME

 
alfa aesar™
B21616ME
500mg Each for $237.05
Description & Specifications

Specifications

Formula Weight 220.19 (211.18 Anhydrous)
Chemical Name or Material 1-Deoxy-1-nitro-L-iditol hemihydrate
Assay 99%
Molecular Formula C6H13NO7·0.5H2O
MDL Number MFCD00191972
CAS 207226-23-1
Melting Point 87° to 89°C
Quantity 500mg