Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Deoxy-1-nitro-D-iditol hemihydrate, 99%

$62.34 - $143.78

Chemical Identifiers

Chemical Name or Material 1-Deoxy-1-nitro-D-iditol hemihydrate
CAS 96613-89-7
Molecular Formula C6H13NO7·0.5H2O
Formula Weight 220.19 (211.18 Anhydrous)
MDL Number MFCD00191971
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB21239MC

 
alfa aesar™
B21239MC
100mg Each for $62.34

AAB21239ME

 
alfa aesar™
B21239ME
500mg Each for $143.78
Description & Specifications

Specifications

Formula Weight 220.19 (211.18 Anhydrous)
Chemical Name or Material 1-Deoxy-1-nitro-D-iditol hemihydrate
Assay 99%
Molecular Formula C6H13NO7·0.5H2O
MDL Number MFCD00191971
CAS 96613-89-7
Melting Point 87° to 89°C
Quantity 100mg