missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Decyne, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 223430050

 View more versions of this product

Catalog No. AC223430050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
Liquid
Clear colorless to light yellow
CH3(CH2)7Cidentical toCH
C10H18
98%
97.5
Glass bottle
Authentic
138.25
5mL
100.0
MFCD00009576
98%
1-Decyne
764-93-2
1.4260 to 1.4280 (20°C, 589nm)

Chemical Identifiers

764-93-2
100.0
C10H18
138.25
1-Decyne, 98%
97.5
98%
CH3(CH2)7Cidentical toCH
MFCD00009576
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-132-8

TSCA : TSCA