Learn More
1-Decylphosphonic acid, 98%, Thermo Scientific Chemicals
$99.27 - $306.79
Chemical Identifiers
CAS | 6874-60-8 |
---|---|
Molecular Formula | C10H23O3P |
Molecular Weight (g/mol) | 222.265 |
MDL Number | MFCD00015830 |
InChI Key | DZQISOJKASMITI-UHFFFAOYSA-N |
Synonym | decanephosphonic acid, phosphonic acid, decyl, 1-decanephosphonic acid, decyl-phosphonic acid, 1-decylphosphonic acid, n-decylphosphonic acid, phosphonic acid, p-decyl, decylphosphonicacid, n-decanephosphonic acid, acmc-1cuhv |
PubChem CID | 81309 |
IUPAC Name | decylphosphonic acid |
SMILES | CCCCCCCCCCP(=O)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1201903
|
Thermo Scientific Chemicals
L1201903 |
1 g |
Each for $99.27
|
|
|||||
AAL1201906
|
Thermo Scientific Chemicals
L1201906 |
5 g |
Each for $306.79
|
|
|||||
Description
1-Decylphosphonic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in stabilizing dye-sensitized solar cell.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Decylphosphonic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used in stabilizing dye-sensitized solar cell.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.
Chemical Identifiers
6874-60-8 | |
222.265 | |
DZQISOJKASMITI-UHFFFAOYSA-N | |
81309 | |
CCCCCCCCCCP(=O)(O)O |
C10H23O3P | |
MFCD00015830 | |
decanephosphonic acid, phosphonic acid, decyl, 1-decanephosphonic acid, decyl-phosphonic acid, 1-decylphosphonic acid, n-decylphosphonic acid, phosphonic acid, p-decyl, decylphosphonicacid, n-decanephosphonic acid, acmc-1cuhv | |
decylphosphonic acid |
Specifications
6874-60-8 | |
C10H23O3P | |
1 g | |
1771572 | |
DZQISOJKASMITI-UHFFFAOYSA-N | |
decylphosphonic acid | |
81309 | |
98% |
100°C to 101°C | |
MFCD00015830 | |
UN3261 | |
decanephosphonic acid, phosphonic acid, decyl, 1-decanephosphonic acid, decyl-phosphonic acid, 1-decylphosphonic acid, n-decylphosphonic acid, phosphonic acid, p-decyl, decylphosphonicacid, n-decanephosphonic acid, acmc-1cuhv | |
CCCCCCCCCCP(=O)(O)O | |
222.265 | |
222.27 | |
1-Decylphosphonic acid |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 229-975-2
RTECSNumber : SZ7525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only