Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Decylboronic acid pinacol ester, 98%

$1,030.62 - $1,030.62

Chemical Identifiers

Chemical Name or Material 1-Decylboronic acid pinacol ester
CAS 141091-38-5
Molecular Formula C16H33BO2
Formula Weight 268.25
MDL Number MFCD12546186
Synonym 2-(n-Decyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3040314

  SDS
alfa aesar™
H3040314
25g Each for $1,030.62
Description & Specifications

Specifications

Formula Weight 268.25
Quantity 25g
Chemical Name or Material 1-Decylboronic acid pinacol ester
Assay 98%
MDL Number MFCD12546186
Molecular Formula C16H33BO2
Synonym 2-(n-Decyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
CAS 141091-38-5