missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Decylboronic acid, 98%, Thermo Scientific Chemicals
$76.01 - $879.40
Chemical Identifiers
CAS | 24464-63-9 |
---|---|
Molecular Formula | C10H23BO2 |
Molecular Weight (g/mol) | 186.102 |
MDL Number | MFCD01074638 |
InChI Key | TUTSEMMBKGBUGD-UHFFFAOYSA-N |
Synonym | n-decylboronic acid, boronic acid, decyl, decylboranic acid, n-decaneboronic acid, n-, boronicacid, b-decyl, 1-decylboronic acid, ksc199m8d |
PubChem CID | 4186635 |
IUPAC Name | decylboronic acid |
SMILES | B(CCCCCCCCCC)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1995703
|
Thermo Scientific Chemicals
L1995703 |
1 g |
Each for $76.01
|
|
|||||
AAL1995706
|
Thermo Scientific Chemicals
L1995706 |
5 g |
Each for $231.71
|
|
|||||
AAL1995714
|
Thermo Scientific Chemicals
L1995714 |
25 g |
Each for $879.40
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24464-63-9 | |
186.102 | |
TUTSEMMBKGBUGD-UHFFFAOYSA-N | |
4186635 | |
B(CCCCCCCCCC)(O)O |
C10H23BO2 | |
MFCD01074638 | |
n-decylboronic acid, boronic acid, decyl, decylboranic acid, n-decaneboronic acid, n-, boronicacid, b-decyl, 1-decylboronic acid, ksc199m8d | |
decylboronic acid |
Specifications
24464-63-9 | |
C10H23BO2 | |
1 g | |
n-decylboronic acid, boronic acid, decyl, decylboranic acid, n-decaneboronic acid, n-, boronicacid, b-decyl, 1-decylboronic acid, ksc199m8d | |
B(CCCCCCCCCC)(O)O | |
186.102 | |
186.1 | |
1-Decylboronic acid |
86°C to 89°C | |
MFCD01074638 | |
1743934 | |
TUTSEMMBKGBUGD-UHFFFAOYSA-N | |
decylboronic acid | |
4186635 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only