Please login to your online account to display your discounted pricing

Alfa Aesar™ (1-Decyl)triphenylphosphonium bromide, 98%

$72.04 - $72.04

Chemical Identifiers

Chemical Name or Material (1-Decyl)triphenylphosphonium bromide
CAS 32339-43-8
Molecular Formula CH3(CH2)9P(C6H5)3Br
Formula Weight 483.48
MDL Number MFCD00051854
Beilstein 4076987
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1102114

  SDS
alfa aesar™
A1102114
25g Each for $72.04
Description & Specifications

Specifications

Formula Weight 483.48
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material (1-Decyl)triphenylphosphonium bromide
Assay 98%
Molecular Formula CH3(CH2)9P(C6H5)3Br
MDL Number MFCD00051854
CAS 32339-43-8
Melting Point ≈90°C
Beilstein 4076987
Quantity 25g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.