Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Decyl isothiocyanate, 96%

Chemical Identifiers

Chemical Name or Material 1-Decyl isothiocyanate
CAS 24540-94-1
Molecular Formula CH3(CH2)9NCS
Formula Weight 199.36g/mol
MDL Number MFCD00041120
UN Number UN1760
Beilstein 6288335
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1216503

 
alfa aesar™
L12165-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1216506

 
alfa aesar™
L12165-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Boiling Point 170° to 171°C (35mmHg)
Quantity 1g
Odor Irritating
Chemical Name or Material 1-Decyl isothiocyanate
CAS 24540-94-1
Assay 96%
Molecular Formula CH3(CH2)9NCS
Formula Weight 199.36g/mol
MDL Number MFCD00041120
UN Number UN1760
Beilstein 6288335
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.