missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Decene, ca. 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC111910025
Specifications
1-Decene | |
49°C | |
Glass bottle | |
0.741 | |
169.0°C to 171.0°C | |
-66.0°C | |
872-05-9 | |
100.0 | |
C10H20 | |
01,III,858 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol | |
CCCCCCCCC=C | |
140.27 | |
CHEBI:87315 | |
Liquid |
0.7410g/mL | |
Authentic | |
1.4200 to 1.4220 | |
1.1 mm2/s (20°C) | |
Colorless | |
2.5L | |
94.0 | |
94% min. (GC) | |
CH3(CH2)7CH=CH2 | |
1-decene, 1-n-decene, alpha-decene, decene, decene-1, decylene, dialene 10, gulftene 10, n-1-decene, n-decylene | |
AFFLGGQVNFXPEV-UHFFFAOYSA-N | |
dec-1-ene | |
13381 | |
140.27 | |
ca. 95% |
Chemical Identifiers
872-05-9 | |
140.27 | |
1-decene, 1-n-decene, alpha-decene, decene, decene-1, decylene, dialene 10, gulftene 10, n-1-decene, n-decylene | |
CHEBI:87315 | |
CCCCCCCCC=C |
C10H20 | |
AFFLGGQVNFXPEV-UHFFFAOYSA-N | |
13381 | |
dec-1-ene |
Safety and Handling
GHS H Statement
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Avoid release to the environment.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Danger
EINECSNumber : 212-819-2