missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Decene, 96%, remainder isomers, Alfa Aesar™

Manufacturer:  Alfa Aesar 31213A3

 View more versions of this product

Catalog No. AA31213A3



Specifications

Specifications

170°C to 171°C
-66°C
1-Decene
49° (120°F)
872-05-9
Remainder isomers
CH3(CH2)7CH=CH2
UN3295
homopolymer hydrogenated oligomers hydrogenated, 1-decene homopolymer hydrogenated, hydrogenated decene homopolymer, 1-decene analytical standard, 1-decene purum gc, polydecene hydrogenated nf, polydecene hydrogenated, hydrogenated polydecene, c10-c16 alpha-olefin, 1-decene gc
CCCCCCCCC=C
140.27
CHEBI:87315
Liquid
Colorless
2kg
0.741g/mL
1.4210
96%
C10H20
MFCD00009577
1737236
AFFLGGQVNFXPEV-UHFFFAOYSA-N
dec-1-ene
13381
140.27
96%

Chemical Identifiers

872-05-9
140.27
AFFLGGQVNFXPEV-UHFFFAOYSA-N
13381
dec-1-ene
C10H20
MFCD00009577
homopolymer hydrogenated oligomers hydrogenated, 1-decene homopolymer hydrogenated, hydrogenated decene homopolymer, 1-decene analytical standard, 1-decene purum gc, polydecene hydrogenated nf, polydecene hydrogenated, hydrogenated polydecene, c10-c16 alpha-olefin, 1-decene gc
CHEBI:87315
CCCCCCCCC=C
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226-H315
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

RTECSNumber : HE2071401

TSCA : TSCA

SDS Documents