missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Decene, 96%, remainder isomers, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 03121336

 View more versions of this product

Catalog No. AA3121336



Description

Description

1-Decene is the only isomer of industrial importance. it is used as a monomer in copolymers and is an intermediate in the production of epoxides, amines, oxo alcohols, synthetic lubricants, synthetic fatty acids, and alkylated aromatics. 1-Decene has been isolated from the leaves and rhizome of the plant Farfugium japonicum and has been detected as the initial product in the microbial degradation of n-decane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Decene
-66°C
0.741 g/mL
49°C (120°F)
Remainder isomers
1.421
MFCD00009577
UN3295
1-decene, decylene, decene, n-1-decene, 1-n-decene, alpha-decene, n-decylene, gulftene 10, dialene 10, decene-1
AFFLGGQVNFXPEV-UHFFFAOYSA-N
dec-1-ene
13381
140.27
Liquid
872-05-9
Colorless
170°C to 171°C
96%
C10H20
CH3(CH2)7CH=CH2
500 g
1737236
Alkenes are virtually insoluble in water,but dissolve in organic solvents.
CCCCCCCCC=C
140.27
CHEBI:87315
96%

Chemical Identifiers

872-05-9
140.27
AFFLGGQVNFXPEV-UHFFFAOYSA-N
13381
dec-1-ene
C10H20
MFCD00009577
1-decene, decylene, decene, n-1-decene, 1-n-decene, alpha-decene, n-decylene, gulftene 10, dialene 10, decene-1
CHEBI:87315
CCCCCCCCC=C
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226-H315
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H226-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 212-819-2

RTECSNumber : HE2071401

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only