Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Decene, 94%

Chemical Identifiers

Chemical Name or Material 1-Decene
CAS 872-05-9
Molecular Formula C10H20
Linear Formula CH3(CH2)7CH=CH2
Formula Weight 140.27
MDL Number MFCD00009577
UN Number UN3295
Beilstein 1737236
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL06800AE

  SDS
alfa aesar™
L06800AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 140.27
Chemical Name or Material 1-Decene
MDL Number MFCD00009577
Assay 94%
Molecular Formula C10H20
RTECS Number HE2071401
TSCA TSCA
Boiling Point 170° to 171°C
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H304-H226-H315
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.
Health Hazard 1 Danger
Melting Point −66°C
Linear Formula CH3(CH2)7CH=CH2
Beilstein 1737236
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
UN Number UN3295
Flash Point 49°C (120°F)
Quantity 100mL
CAS 872-05-9

Applications
1-Decene exists as an alpha olefin, it is used as a monomer in copolymers. It is used as an intermediate in the production of epoxides, amines, oxo alcohols, synthetic lubricants, synthetic fatty acids, and alkylated aromatics.

Solubility
Miscible in Ether, Ethanol. Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Safety and Handling

Danger

GHS P Statement
P210-P280-P301+P310-P303+P361+P353-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H304-H226-H315
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Causes skin irritation.