Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Decanesulfonyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 1-Decanesulfonyl chloride
CAS 61652-81-1
Molecular Formula CH3(CH2)9SO2Cl
Formula Weight 240.79
MDL Number MFCD00051746
UN Number UN3261
Beilstein 1771852
Synonym 1-Decylsulfonyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0664209

 
alfa aesar™
L0664209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0664218

 
alfa aesar™
L0664218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 240.79
Molecular Formula CH3(CH2)9SO2Cl
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-Decanesulfonyl chloride
TSCA TSCA
Assay 98%
MDL Number MFCD00051746
CAS 61652-81-1
UN Number UN3261
Melting Point 30° to 34°C
Quantity 10g
Synonym 1-Decylsulfonyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Beilstein 1771852
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Applications
1-Decanesulfonyl chloride is used as pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.