missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Decanesulfonic Acid, Sodium Salt, Anhydrous, HPLC Grade, EMD Millipore

Manufacturer:  Millipore Sigma DX01013

 View more versions of this product

Catalog No. MDX01013



Specifications

Specifications

1-Decanesulfonic Acid, Sodium Salt, Anhydrous
>3°C
For Synthesis
HPLC
sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
CCCCCCCCCCS(=O)(=O)[O-].[Na+]
244.325
244.33
White
100g
13419-61-9
C10H21NaO3S
AIMUHNZKNFEZSN-UHFFFAOYSA-M
sodium;decane-1-sulfonate
2724181
Crystalline Solid

Chemical Identifiers

13419-61-9
244.325
sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
sodium;decane-1-sulfonate
C10H21NaO3S
AIMUHNZKNFEZSN-UHFFFAOYSA-M
2724181
CCCCCCCCCCS(=O)(=O)[O-].[Na+]
Safety and Handling

Safety and Handling

Recommended Storage : Store at +15°C to +25°C

Documents