Please login to your online account to display your discounted pricing

1-Decanesulfonic Acid, Sodium Salt, Anhydrous, HPLC Grade, EMD Millipore

$375.63 - $1,143.91

Chemical Identifiers

Chemical Name or Material 1-Decanesulfonic Acid, Sodium Salt, Anhydrous
Name Note For Synthesis
CAS 13419-61-9
Purity Grade HPLC Grade
Physical Form Crystalline Solid
Molecular Formula CH3(CH2)9SO3Na
Formula Weight 244.33
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MDX01013

 
emd millipore corporation
DX01013
100g Each for $1,143.91

MDX01012

 
emd millipore corporation
DX01012
25g Each for $375.63
Description & Specifications

Specifications

Purity Grade HPLC Grade
Physical Form Crystalline Solid
Formula Weight 244.33
Color White
Quantity 100g
Melting Point >3°C
Chemical Name or Material 1-Decanesulfonic Acid, Sodium Salt, Anhydrous
Name Note For Synthesis
Molecular Formula CH3(CH2)9SO3Na
CAS 13419-61-9
Recommended Storage Store at +15°C to +25°C
Safety and Handling

Store at +15°C to +25°C