Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(Cyclopropylmethyl)piperazine dihydrochloride, 97%

Chemical Identifiers

Chemical Name or Material 1-(Cyclopropylmethyl)piperazine dihydrochloride
CAS 373608-42-5
Formula Weight 213.15
MDL Number MFCD08544420
Beilstein 6420036
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6687003

 
alfa aesar™
H6687003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6687006

 
alfa aesar™
H6687006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.15
MDL Number MFCD08544420
CAS 373608-42-5
Assay 97%
Physical Form Solid
Beilstein 6420036
Quantity 1g
Chemical Name or Material 1-(Cyclopropylmethyl)piperazine dihydrochloride