missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(Cyclopropylcarbonyl)piperazine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5521903
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1-(Cyclopropylcarbonyl)piperazine | |
Brown to Colorless | |
120°C (15 mmHg) | |
C8H15N2O | |
MFCD06369645 | |
1-cyclopropylcarbonyl piperazine, cyclopropyl piperazin-1-yl methanone, 1-cyclopropanecarbonylpiperazine, cyclopropyl-piperazin-1-yl-methanone, 1-cyclopropanecarbonyl piperazine, piperazine, 1-cyclopropylcarbonyl, piperazinocyclopropyl ketone, cyclopropyl piperazinyl ketone, 1-cyclopropylcarbonylpiperazine, 4-cyclopropanecarbonyl-piperazine | |
O=C(C1CC1)N1CC[NH2+]CC1 | |
155.22 | |
154.21 | |
Liquid |
59878-57-8 | |
1.087 g/mL | |
>110°C (230°F) | |
1.524 | |
1 g | |
KIALFUYSJAAJSU-UHFFFAOYSA-O | |
cyclopropyl(piperazin-1-yl)methanone | |
2064235 | |
97% |
Chemical Identifiers
59878-57-8 | |
155.22 | |
KIALFUYSJAAJSU-UHFFFAOYSA-O | |
2064235 | |
O=C(C1CC1)N1CC[NH2+]CC1 |
C8H15N2O | |
MFCD06369645 | |
1-cyclopropylcarbonyl piperazine, cyclopropyl piperazin-1-yl methanone, 1-cyclopropanecarbonylpiperazine, cyclopropyl-piperazin-1-yl-methanone, 1-cyclopropanecarbonyl piperazine, piperazine, 1-cyclopropylcarbonyl, piperazinocyclopropyl ketone, cyclopropyl piperazinyl ketone, 1-cyclopropylcarbonylpiperazine, 4-cyclopropanecarbonyl-piperazine | |
cyclopropyl(piperazin-1-yl)methanone |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only