Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclopropyl-4-piperidone, 98%

$83.18 - $714.20

Chemical Identifiers

Chemical Name or Material 1-Cyclopropyl-4-piperidone
CAS 62813-01-8
Molecular Formula C8H13NO
Formula Weight 139.2
MDL Number MFCD07374375
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2749503

  SDS
alfa aesar™
H2749503
1g Each for $83.18

AAH2749506

  SDS
alfa aesar™
H2749506
5g Each for $236.57

AAH2749514

  SDS
alfa aesar™
H2749514
25g Each for $714.20
Description & Specifications

Specifications

Formula Weight 139.2
Quantity 1g
Assay 98%
Chemical Name or Material 1-Cyclopropyl-4-piperidone
Molecular Formula C8H13NO
MDL Number MFCD07374375
CAS 62813-01-8