Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-Cyclopentyl-2,2-dimethyl-1-propanol, 98%

$150.07 - $629.82

Chemical Identifiers

Chemical Name or Material 1-Cyclopentyl-2,2-dimethyl-1-propanol
CAS 337966-85-5
Molecular Formula C10H20O
Formula Weight 156.27
MDL Number MFCD00060800
Synonym tert-Butyl cyclopentyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2031606

  SDS
alfa aesar™
B2031606
5g Each for $150.07

AAB2031614

  SDS
alfa aesar™
B2031614
25g Each for $629.82
Description & Specifications

Specifications

Formula Weight 156.27
Molecular Formula C10H20O
Assay 98%
CAS 337966-85-5
Melting Point 48° to 50°C
Synonym tert-Butyl cyclopentyl carbinol
MDL Number MFCD00060800
Quantity 5g
Health Hazard 1 Warning
Chemical Name or Material 1-Cyclopentyl-2,2-dimethyl-1-propanol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.