missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC444750010
Specifications
1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester | |
38°C to 40°C | |
1233526-60-7 | |
C14H23BN2O2 | |
OCBNABJVDPSMQY-UHFFFAOYSA-N | |
1-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole | |
56776730 | |
97% |
Glass Bottle | |
1g | |
96% min. (GC) | |
1-cyclopentyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCCC3 | |
262.16 | |
262.16 |
Chemical Identifiers
1233526-60-7 | |
262.16 | |
1-cyclopentyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
1-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole |
C14H23BN2O2 | |
OCBNABJVDPSMQY-UHFFFAOYSA-N | |
56776730 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCCC3 |
Safety and Handling
GHS Signal Word: Warning