Please login to your online account to display your discounted pricing

1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Acros Organics

Chemical Identifiers

Chemical Name or Material 1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester
Assay Percent Range 97%
Molecular Formula C14H23BN2O2
Formula Weight 262.16
Synonym 1-Cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC444750010

  SDS
thermo fisher scientific
AC444750010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC444750050

  SDS
thermo fisher scientific
AC444750050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 262.16
Quantity 1g
Chemical Name or Material 1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester
CAS 1233526-60-7
Synonym 1-Cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular Formula C14H23BN2O2
Assay Percent Range 97%