missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC444750050

 View more versions of this product

Catalog No. AC444750050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester
5g
96% min. (GC)
1-cyclopentyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCCC3
262.16
262.16
38°C to 40°C
1233526-60-7
C14H23BN2O2
OCBNABJVDPSMQY-UHFFFAOYSA-N
1-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
56776730
97%

Chemical Identifiers

1233526-60-7
262.16
1-cyclopentyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
1-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
C14H23BN2O2
OCBNABJVDPSMQY-UHFFFAOYSA-N
56776730
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCCC3
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents